Ethyl-2-piperidinecarboxylate 97%
Supplier: Apollo Scientific
Synonyms: Ethyl 2-Piperidinecarboxylate, Ethyl pipecolinate
OR5106-100G OR5106-25G
APOSOR5106-100GEA 77.4 EUR
APOSOR5106-100G APOSOR5106-25G
Ethyl-2-piperidinecarboxylate 97%
Ethyl 2-piperidinecarboxylate
Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Boiling Pt: 216…217 °C
Density: 1.006 g/cm³ (25 °C)
Flash Pt: 46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005980
CAS Number: 15862-72-3