Isoquinoline
Supplier: Apollo Scientific
Danger
OR5109-100G OR5109-500G OR5109-25G
APOSOR5109-100GEA 36.4 EUR
APOSOR5109-100G APOSOR5109-500G APOSOR5109-25G
Isoquinoline
Isoquinoline
Formula: C₉H₇N
MW: 129.16 g/mol
Boiling Pt: 242…243 °C
Melting Pt: 26…28 °C
Density: 1.099 g/cm³ (25 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00006898
CAS Number: 119-65-3
EINECS: 204-341-8
UN: 2811
ADR: 6.1,III