2-Amino-4-chloropyridine 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Chloro-2-pyridylamine, 4-Chloropyridin-2-amine
OR5114-25G
OR5114-5G
OR5114-100G
APOSOR5114-25GEA
26.1
EUR
APOSOR5114-25G
APOSOR5114-5G
APOSOR5114-100G
2-Amino-4-chloropyridine 98%
2-Amino-4-chloropyridine
Formula:
C₅H₅ClN₂ MW: 128.56 g/mol Melting Pt: 129…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD04113820 CAS Number: 19798-80-2 |