2-Nitro-m-cresol
Supplier: Apollo Scientific
Warning
Synonyms: 2-Nitro-3-methylphenol, 3-Methyl-2-nitrophenol
OR5116-10G OR5116-50G OR5116-25G OR5116-100G
APOSOR5116-10GEA 21.9 EUR
APOSOR5116-10G APOSOR5116-50G APOSOR5116-25G APOSOR5116-100G
2-Nitro-m-cresol
2-Nitro-m-cresol
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 106…108 °C/9.5mmHg
Melting Pt: 35…39 °C
Flash Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007125
CAS Number: 4920-77-8
EINECS: 225-546-9