1-(4-Aminophenyl)-4-(4-methoxyphenyl)piperazine
Supplier: Apollo Scientific
Synonyms: 4-[4-(4-Methoxyphenyl)-1-piperazinyl]aniline, 4-(4-(4-Methoxyphenyl)piperazin-1-yl)aniline
OR5129-25G OR5129-5G
APOSOR5129-25GEA 601 EUR
APOSOR5129-25G APOSOR5129-5G
1-(4-Aminophenyl)-4-(4-methoxyphenyl)piperazine
1-(4-Aminophenyl)-4-(4-methoxyphenyl)piperazine
Formula: C₁₇H₂₁N₃O
MW: 283.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02071496
CAS Number: 74852-62-3