cis-2,6-Dimethylpiperazine 98+%
Supplier: Apollo Scientific
Danger
OR5153-100G OR5153-25G OR5153-5G
APOSOR5153-100GEA 129 EUR
APOSOR5153-100G APOSOR5153-25G APOSOR5153-5G
cis-2,6-Dimethylpiperazine 98+%
cis-2,6-Dimethylpiperazine
Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 162 °C
Melting Pt: 108…111 °C
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD07772435
CAS Number: 21655-48-1
EINECS: 203-588-9
UN: 1325
ADR: 4.1,III