4-(Methylsulphonyl)aniline
Supplier: Apollo Scientific
Warning
OR5161-100G OR5161-25G
APOSOR5161-100GEA 48 EUR
APOSOR5161-100G APOSOR5161-25G
4-(Methylsulphonyl)aniline
4-(Methylsulphonyl)aniline
Formula: C₇H₉NO₂S
MW: 171.21 g/mol
Melting Pt: 133…138 °C
Density: 1.44 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00025353
CAS Number: 5470-49-5
EINECS: 226-803-8