(S)-(-)-Epoxystyrene 98%

Supplier: Apollo Scientific

Synonyms: (S)-Styrene oxide

OR5172-100G OR5172-25G OR5172-5G
APOSOR5172-100GEA 455 EUR
APOSOR5172-100G APOSOR5172-25G APOSOR5172-5G
(S)-(-)-Epoxystyrene 98%
(S)-(-)-Epoxystyrene
Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 192…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00064310
CAS Number: 20780-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR