2-Chlorobenzoic acid
Supplier: Apollo Scientific
Warning
OR5177-1KG OR5177-500G
APOSOR5177-1KGEA 31.9 EUR
APOSOR5177-1KG APOSOR5177-500G
2-Chlorobenzoic acid
2-Chlorobenzoic acid
Formula: C₇H₅ClO₂
Boiling Pt: 285 °C
Melting Pt: 138…143 °C
Density: 1.544 g/cm³ (25 °C)
Flash Pt: 173 °C
Storage Temperature: Ambient
MDL Number: MFCD00002412
CAS Number: 118-91-2
EINECS: 204-285-4