3-Bromo-4-pyridone 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-4-hydroxypyridine, 3-Bromo-4-pyridinol
OR51772-10G OR51772-5G
APOSOR51772-10GEA 78.8 EUR
APOSOR51772-10G APOSOR51772-5G
3-Bromo-4-pyridone 97%
3-Bromo-4-pyridone
Formula: C₅H₄BrNO
MW: 174.00 g/mol
Melting Pt: 225…230 °C
Storage Temperature: Ambient
MDL Number: MFCD01646107
CAS Number: 36953-41-0