6-Nitro-m-toluidine
Supplier: Apollo Scientific
Danger
Synonyms: 6-Nitro-3-methylaniline, 3-Amino-4-nitrotoluene, 5-Methyl-2-nitroaniline
OR51790-25G OR51790-5G
APOSOR51790-25GEA 83.2 EUR
APOSOR51790-25G APOSOR51790-5G
6-Nitro-m-toluidine
6-Nitro-m-toluidine
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Melting Pt: 110…111 °C
Storage Temperature: Ambient
MDL Number: MFCD00007814
CAS Number: 578-46-1
EINECS: 209-423-7
UN: 2660
ADR: 6.1 III