(R)-(-)-3-Isopropyl-2,5-piperazinedione
Supplier: Apollo Scientific
Warning
Synonyms: (R)-3-Isopropylpiperazine-2,5-dione
OR51793-5G
APOSOR51793-5GEA 120 EUR
APOSOR51793-5G
(R)-(-)-3-Isopropyl-2,5-piperazinedione
(R)-(-)-3-Isopropyl-2,5-piperazinedione
Formula: C₇H₁₂N₂O₂
MW: 156.18 g/mol
Melting Pt: 256…262 °C
Storage Temperature: Ambient
MDL Number: MFCD00210060
CAS Number: 143673-66-9