5-Bromo-2-chloropyridine 98%
Supplier: Apollo Scientific
Warning
OR5181-100G OR5181-25G
APOSOR5181-100GEA 68.3 EUR
APOSOR5181-100G APOSOR5181-25G
5-Bromo-2-chloropyridine 98%
5-Bromo-2-chloropyridine
Formula: C₅H₃BrClN
MW: 192.44 g/mol
Melting Pt: 65…72 °C
Storage Temperature: Ambient
MDL Number: MFCD01318951
CAS Number: 53939-30-3