2,4-Dichloro-5-methoxypyrimidine
Supplier: Apollo Scientific
Warning
OR5184-1G OR5184-25G
APOSOR5184-1GEA 21.9 EUR
APOSOR5184-1G APOSOR5184-25G
2,4-Dichloro-5-methoxypyrimidine
2,4-Dichloro-5-methoxypyrimidine
Formula: C₅H₄Cl₂N₂O
Melting Pt: 66…70 °C
Storage Temperature: Ambient
MDL Number: MFCD02685670
CAS Number: 19646-07-2