2-Bromo-4-methoxybenzoic acid 98%
Supplier: Apollo Scientific
Warning
OR51862-1G OR51862-5G OR51862-25G
APOSOR51862-1GEA 21.9 EUR
APOSOR51862-1G APOSOR51862-5G APOSOR51862-25G
2-Bromo-4-methoxybenzoic acid 98%
2-Bromo-4-methoxybenzoic acid
Formula: C₈H₇BrO₃
MW: 231.0434 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11044843
CAS Number: 74317-85-4