1-Chloro-2-iodoethane
Supplier: Apollo Scientific
Warning
OR51875-1G OR51875-5G
APOSOR51875-1GEA 71.2 EUR
APOSOR51875-1G APOSOR51875-5G
1-Chloro-2-iodoethane
1-Chloro-2-iodoethane
Formula: C₂H₄ClI
MW: 190.41 g/mol
Boiling Pt: 61…62 °C
Melting Pt: -15.6
Density: 2.088 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041453
CAS Number: 624-70-4