N-Benzylhydroxylamine hydrochloride 97%
Supplier: Apollo Scientific
Warning
OR51891-25G OR51891-5G OR51891-100G
APOSOR51891-25GEA 21.9 EUR
APOSOR51891-25G APOSOR51891-5G APOSOR51891-100G
N-Benzylhydroxylamine hydrochloride 97%
N-Benzylhydroxylamine hydrochloride
Formula: C₇H₁₀ClNO
MW: 159.6134 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00043377
CAS Number: 29601-98-7