Ethyl-2-methylacetoacetate
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 2-methyl-3-oxobutanoate
OR51898-100G OR51898-25G
APOSOR51898-100GEA 39.3 EUR
APOSOR51898-100G APOSOR51898-25G
Ethyl-2-methylacetoacetate
Ethyl 2-methylacetoacetate
Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 187 °C
Melting Pt: -45
Density: 1.019 g/cm³ (25 °C)
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00009164
CAS Number: 609-14-3
EINECS: 210-179-9