2,3-Dihydroxybenzaldehyde 98%
Supplier: Apollo Scientific
Warning
OR51920-25G OR51920-100G
APOSOR51920-25GEA 66.7 EUR
APOSOR51920-25G APOSOR51920-100G
2,3-Dihydroxybenzaldehyde 98%
2,3-Dihydroxybenzaldehyde
Formula: C₇H₆O₃
MW: 138.12 g/mol
Melting Pt: 104…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00003324
CAS Number: 24677-78-9
EINECS: 246-398-1