2-Piperidone 97%
Supplier: Apollo Scientific
Warning
Synonyms: δ-Valerolactame, delta-Valerolactam, Piperidin-2-one
OR5194-100G OR5194-25G OR5194-500G
APOSOR5194-100GEA 56.7 EUR
APOSOR5194-100G APOSOR5194-25G APOSOR5194-500G
2-Piperidone 97%
2-Piperidone
Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 256 °C
Melting Pt: 38…40 °C
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00006037
CAS Number: 675-20-7