2,4,6-Tris(3',5'-di-tert-butyl-4'-hydroxybenzyl)mesitylene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1,3,5-Trimethyl-2,4,6-tris(3,5-di-tert-butyl-4-hydroxybenzyl)benzene, 4,4',4''-((2,4,6-Trimethylbenzene-1,3,5-triyl)tris(methylene))tris(2,6-di-tert-butylphenol)
OR51956-100G OR51956-500G OR51956-1KG
APOSOR51956-100GEA 23.2 EUR
APOSOR51956-100G APOSOR51956-500G APOSOR51956-1KG
2,4,6-Tris(3',5'-di-tert-butyl-4'-hydroxybenzyl)mesitylene 97%
2,4,6-Tris(3',5'-di-tert-butyl-4'-hydroxybenzyl)mesitylene
Formula: C₅₄H₇₈O₃
MW: 775.22 g/mol
Melting Pt: 248…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00026284
CAS Number: 1709-70-2
EINECS: 216-971-0