2-Bromo-5-nitrophenol
Supplier: Apollo Scientific
Warning
OR51974-1G OR51974-25G OR51974-5G
APOSOR51974-1GEA 21.9 EUR
APOSOR51974-1G APOSOR51974-25G APOSOR51974-5G
2-Bromo-5-nitrophenol
2-Bromo-5-nitrophenol
Formula: C₆H₄BrNO₃
MW: 218.0100 g/mol
Melting Pt: 116…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00466299
CAS Number: 52427-05-1
UN: 3077
ADR: 9+EHS III