Ethyl-3-aminobenzoate
Supplier: Apollo Scientific
Warning
OR52047-100G OR52047-250G OR52047-500G
APOSOR52047-100GEA 96.3 EUR
APOSOR52047-100G APOSOR52047-250G APOSOR52047-500G
Ethyl-3-aminobenzoate
Ethyl 3-aminobenzoate
Formula: C₉H₁₁NO₂
Boiling Pt: 172…175 °C/13mmHg
Density: 1.107 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007794
CAS Number: 582-33-2
EINECS: 209-482-9