(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol 97%
Supplier: Apollo Scientific
Warning
Synonyms: (1S,2S,3R,5S)-(+)-2,3-Pinanediol, (1S,2S,3R,5S)-(+)-Pinanediol
OR52059-100G OR52059-25G OR52059-5G
APOSOR52059-100GEA 235 EUR
APOSOR52059-100G APOSOR52059-25G APOSOR52059-5G
(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol 97%
(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol
Formula: C₁₀H₁₈O₂
MW: 170.25 g/mol
Melting Pt: 52…60 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077851
CAS Number: 18680-27-8