Glyoxylic acid monohydrate 97%
Supplier: Apollo Scientific
Danger
Synonyms: Formylformic acid monohydrate, Oxoacetic acid monohydrate, 2-Oxoethanoic acid monohydrate
OR52099-100G OR52099-1KG OR52099-500G
APOSOR52099-100GEA 60.9 EUR
APOSOR52099-100G APOSOR52099-1KG APOSOR52099-500G
Glyoxylic acid monohydrate 97%
Glyoxylic acid monohydrate
Formula: C₂H₂O₃·1H₂O
MW: 92.05 g/mol
Boiling Pt: 100 °C
Melting Pt: 49…52 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00127974
CAS Number: 563-96-2
EINECS: 206-058-5
UN: 3261
ADR: 8,III