imidazo[1,2-a]pyridine-7-carbohydrazide
Supplier: Apollo Scientific
Warning
Synonyms: Imidazo[1,2-a]pyridine-7-carbohydrazide
OR52104-1G OR52104-250MG
APOSOR52104-1GEA 248 EUR
APOSOR52104-1G APOSOR52104-250MG
imidazo[1,2-a]pyridine-7-carbohydrazide
imidazo[1,2-a]pyridine-7-carbohydrazide
Formula: C₈H₈N₄O
MW: 176.179 g/mol
Melting Pt: 258…261 °C
Storage Temperature: Ambient
MDL Number: MFCD13183226
CAS Number: 421595-78-0