6-Bromobenzothiazole 97%
Supplier: Apollo Scientific
Warning
Synonyms: 6-Bromobenzo[d]thiazole
OR52121-5G
APOSOR52121-5GEA 69.6 EUR
APOSOR52121-5G
6-Bromobenzothiazole 97%
6-Bromobenzothiazole
Formula: C₇H₄BrNS
MW: 214.09 g/mol
Melting Pt: 52…54 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115372
CAS Number: 53218-26-1
UN: 2811
ADR: 6.1,III