3,6-Dihydro-2H-pyran-4-boronic acid pinacol ester 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3,6-Dihydro-2H-pyran-4-boronic acid Pinacol ester, 2-(3,6-Dihydro-2H-pyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,6-Dihydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-pyran, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyran
OR52123-1G OR52123-25G OR52123-5G
APOSOR52123-1GEA 34.8 EUR
APOSOR52123-1G APOSOR52123-25G APOSOR52123-5G
3,6-Dihydro-2H-pyran-4-boronic acid pinacol ester 97%
3,6-Dihydro-2H-pyran-4-boronic acid pinacol ester
Formula: C₁₁H₁₉BO₃
MW: 210.08 g/mol
Melting Pt: 63…67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11052631
CAS Number: 287944-16-5