3,4-Dihydroxybenzoic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Protocatechuic acid
OR52125-100G
OR52125-25G
OR52125-500G
APOSOR52125-100GEA
96
EUR
APOSOR52125-100G
APOSOR52125-25G
APOSOR52125-500G
3,4-Dihydroxybenzoic acid
3,4-Dihydroxybenzoic acid
Formula:
C₇H₆O₄ MW: 154.12 g/mol Melting Pt: 197…200 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002509 CAS Number: 99-50-3 EINECS: 202-760-0 Merck Index: 12,08080 |