(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine 95%
Supplier: Apollo Scientific
Danger
Synonyms: (1R,2R)-(-)-1,2-Bis(methylamino)cyclohexane, (1R,2R)-N,N'-Dimethyl-1,2-cyclohexanediamine
OR52130-5G OR52130-25G OR52130-1G
APOSOR52130-5GEA 24.8 EUR
APOSOR52130-5G APOSOR52130-25G APOSOR52130-1G
(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine 95%
(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine
Formula: C₈H₁₈N₂
MW: 142.25 g/mol
Boiling Pt: 78…80 °C/18mmHg
Melting Pt: 36…44 °C
Density: 0.902 g/cm³ (25 °C)
Flash Pt: 74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00671527
CAS Number: 68737-65-5
UN: 3259
ADR: 8 III