3-Methoxypyrocatechol
Supplier: Apollo Scientific
Warning
Synonyms: 2,3-Dihydroxyanisole, 3-Methoxycatechol, 3-Methoxybenzene-1,2-diol
OR52178-25G OR52178-100G
APOSOR52178-25GEA 97.3 EUR
APOSOR52178-25G APOSOR52178-100G
3-Methoxypyrocatechol
3-Methoxypyrocatechol
Formula: C₇H₈O₃
MW: 140.14 g/mol
Boiling Pt: 146…147 °C
Melting Pt: 38…43 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002191
CAS Number: 934-00-9
EINECS: 213-276-4