Triethyl borate
Supplier: Apollo Scientific
Danger
OR52253-100ML OR52253-250ML
APOSOR52253-100MLEA 65.5 EUR
APOSOR52253-100ML APOSOR52253-250ML
Triethyl borate
Triethyl borate
Formula: C₆H₁₅BO₃
MW: 145.99 g/mol
Boiling Pt: 117…118 °C
Melting Pt: –85 °C
Density: 0.858 g/cm³ (25 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00012846
CAS Number: 150-46-9
EINECS: 205-760-9
UN: 1176
ADR: 3 II