1,3-Dibromo-7-tert-butylpyrene 97%
Supplier: Apollo Scientific
OR52284-1G OR52284-5G
APOSOR52284-1GEA 85.7 EUR
APOSOR52284-1G APOSOR52284-5G
1,3-Dibromo-7-tert-butylpyrene 97%
1,3-Dibromo-7-tert-butylpyrene
Formula: C₂₀H₁₆Br₂
MW: 416.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22581440
CAS Number: 1005771-04-9