(3,4-Dimethoxyphenyl)acetonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 3,4-Dimethoxyphenylacetonitrile, Homoveratronitrile, 3,4-Dimethoxybenzyl cyanide
OR52311-100G OR52311-500G
APOSOR52311-100GEA 69.8 EUR
APOSOR52311-100G APOSOR52311-500G
(3,4-Dimethoxyphenyl)acetonitrile
(3,4-Dimethoxyphenyl)acetonitrile
Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Boiling Pt: 171…178 °C/10mmHg
Melting Pt: 62…65 °C
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00001911
CAS Number: 93-17-4
EINECS: 202-225-1