2-Methoxyphenothiazine 98%
Supplier: Apollo Scientific
Warning
OR52313-25G OR52313-100G
APOSOR52313-25GEA 29 EUR
APOSOR52313-25G APOSOR52313-100G
2-Methoxyphenothiazine 98%
2-Methoxyphenothiazine
Formula: C₁₃H₁₁NOS
MW: 229.30 g/mol
Melting Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00037838
CAS Number: 1771-18-2