Boc-b-Alanine 98%
Supplier: Apollo Scientific
Warning
Synonyms: N-Boc-beta-alanine, Boc-β-Ala-OH, BOC-beta-alanine
OR52389-25G OR52389-500G OR52389-100G OR52389-1KG
APOSOR52389-25GEA 0 EUR
APOSOR52389-25G APOSOR52389-500G APOSOR52389-100G APOSOR52389-1KG
Boc-b-Alanine 98%
Boc-b-Alanine
Formula: C₈H₁₅NO₄
Boiling Pt: 138…142 °C/0.3mmHg
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00037291
CAS Number: 3303-84-2
EINECS: 221-979-2