2-Naphthoic acid 98%
Supplier: Apollo Scientific
Warning
OR52474-100G OR52474-500G
APOSOR52474-100GEA 69.8 EUR
APOSOR52474-100G APOSOR52474-500G
2-Naphthoic acid 98%
2-Naphthoic acid
Formula: C₁₁H₈O₂
MW: 172.18 g/mol
Melting Pt: 185…187 °C
Density: 1.08 g/cm³ (25 °C)
Flash Pt: 205 °C
Storage Temperature: Ambient
MDL Number: MFCD00004101
CAS Number: 93-09-4
EINECS: 202-217-8