2-Butyne 98%
Supplier: Apollo Scientific
Danger
OR52506-25G OR52506-5G
APOSOR52506-25GEA 277 EUR
APOSOR52506-25G APOSOR52506-5G
2-Butyne 98%
2-Butyne
Formula: C₄H₆
Boiling Pt: 27 °C
Melting Pt: –32 °C
Density: 0.69 g/cm³ (25 °C)
Flash Pt: –25 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009275
CAS Number: 503-17-3
EINECS: 207-962-2
UN: 1144
ADR: 3,I