2-(2,4-Dichlorophenoxy)propionic acid hydrazide 97%
Supplier: Apollo Scientific
OR5251-10G OR5251-25G OR5251-50G
APOSOR5251-10GEA 129 EUR
APOSOR5251-10G APOSOR5251-25G APOSOR5251-50G
2-(2,4-Dichlorophenoxy)propionic acid hydrazide 97%
2-(2,4-Dichlorophenoxy)propionic acid hydrazide
Formula: C₉H₁₀Cl₂N₂O₂
MW: 249.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00204152
CAS Number: 15253-89-1