5,7-Dichloropyrazolo[1,5-a]pyrimidine
Supplier: Apollo Scientific
Warning
OR52545-25G OR52545-5G
APOSOR52545-25GEA 319 EUR
APOSOR52545-25G APOSOR52545-5G
5,7-Dichloropyrazolo[1,5-a]pyrimidine
5,7-Dichloropyrazolo[1,5-a]pyrimidine
Formula: C₆H₃Cl₂N₃
MW: 188.0141 g/mol
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD08273918
CAS Number: 57489-77-7