2,5-Dichloroaniline 99%
Supplier: Apollo Scientific
Danger
OR5255-1KG OR5255-250G OR5255-5KG
APOSOR5255-1KGEA 24.8 EUR
APOSOR5255-1KG APOSOR5255-250G APOSOR5255-5KG
2,5-Dichloroaniline 99%
2,5-Dichloroaniline
Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Boiling Pt: 244…251 °C
Melting Pt: 47…50 °C
Density: 1.54 g/cm³ (25 °C)
Flash Pt: 139 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007667
CAS Number: 95-82-9
EINECS: 202-455-2
UN: 3442
ADR: 6.1,II