5-Chloro-3-nitropyrazolo[1,5-a]pyrimidine 97%
Supplier: Apollo Scientific
Warning
OR52568-1G OR52568-5G OR52568-10G
APOSOR52568-1GEA 84.1 EUR
APOSOR52568-1G APOSOR52568-5G APOSOR52568-10G
5-Chloro-3-nitropyrazolo[1,5-a]pyrimidine 97%
5-Chloro-3-nitropyrazolo[1,5-a]pyrimidine
Formula: C₆H₃ClN₄O₂
MW: 198.57 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22415211
CAS Number: 1363380-51-1