2-Amino-5-nitrophenol
Supplier: Apollo Scientific
Warning
Synonyms: 2-Hydroxy-4-nitroaniline
OR5257-100G OR5257-25G
APOSOR5257-100GEA 152 EUR
APOSOR5257-100G APOSOR5257-25G
2-Amino-5-nitrophenol
2-Amino-5-nitrophenol
Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 198…200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007692
CAS Number: 121-88-0
EINECS: 204-503-8