2,5-Diaminotoluene 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Methyl-1,4-phenylenediamine
OR52648-25G OR52648-5G OR52648-100G
APOSOR52648-25GEA 151 EUR
APOSOR52648-25G APOSOR52648-5G APOSOR52648-100G
2,5-Diaminotoluene 98%
2,5-Diaminotoluene
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Melting Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00035779
CAS Number: 95-70-5
EINECS: 202-442-1
UN: 2811
ADR: 6.1,III