3-Amino-N-isopropylbenzamide
Supplier: Apollo Scientific
Warning
OR52660-1G OR52660-5G
APOSOR52660-1GEA 78.3 EUR
APOSOR52660-1G APOSOR52660-5G
3-Amino-N-isopropylbenzamide
3-Amino-N-isopropylbenzamide
Formula: C₁₀H₁₄N₂O
MW: 178.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09045316
CAS Number: 81882-62-4
EINECS: 279-842-8