3-Amino-N-cyclohexylbenzamide
Supplier: Apollo Scientific
Warning
OR52661-1G OR52661-5G
APOSOR52661-1GEA 152 EUR
APOSOR52661-1G APOSOR52661-5G
3-Amino-N-cyclohexylbenzamide
3-Amino-N-cyclohexylbenzamide
Formula: C₁₃H₁₈N₂O
MW: 218.3 g/mol
Melting Pt: 146 °C
Storage Temperature: Ambient
MDL Number: MFCD08721753
CAS Number: 77201-15-1