Dibenzofuran
Supplier: Apollo Scientific
Warning
Synonyms: Dibenzo[b,d]furan
OR5267-100G OR5267-25G OR5267-50G
APOSOR5267-100GEA 43.5 EUR
APOSOR5267-100G APOSOR5267-25G APOSOR5267-50G
Dibenzofuran
Dibenzofuran
Formula: C₁₂H₈O
Boiling Pt: 287…289 °C
Melting Pt: 80…82 °C
Density: 1.089 g/cm³ (25 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00004968
CAS Number: 132-64-9
EINECS: 205-071-3
UN: 3077
ADR: 9+EHS,III