(S)-(-)-α-Ethylbenzylamine
Supplier: Apollo Scientific
Danger
Synonyms: (S)-(-)-1-Phenylpropylamine
OR52676-25G OR52676-5G
APOSOR52676-25GEA 379 EUR
APOSOR52676-25G APOSOR52676-5G
(S)-(-)-α-Ethylbenzylamine
(S)-(-)-α-Ethylbenzylamine
Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 205 °C
Melting Pt: –69 °C
Density: 0.94 g/cm³ (25 °C)
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00082356
CAS Number: 3789-59-1
UN: 2735
ADR: 8,II