4-Bromo-2-(4-morpholino)pyridine 98%
Supplier: Apollo Scientific
Synonyms: 4-(4-Bromopyridin-2-yl)morpholine, 4-(4-Bromo-2-pyridyl)morpholine
OR52762-1G OR52762-250MG OR52762-5G
APOSOR52762-1GEA 249 EUR
APOSOR52762-1G APOSOR52762-250MG APOSOR52762-5G
4-Bromo-2-(4-morpholino)pyridine 98%
4-Bromo-2-(4-morpholino)pyridine
Formula: C₉H₁₁BrN₂O
MW: 243.1004 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13195616
CAS Number: 1040377-12-5