1-Phenylethyl mercaptan 98%
Supplier: Apollo Scientific
OR52781-10G OR52781-1G
APOSOR52781-10GEA 292 EUR
APOSOR52781-10G APOSOR52781-1G
1-Phenylethyl mercaptan 98%
1-Phenylethyl mercaptan
Formula: C₈H₁₀S
MW: 138.23 g/mol
Boiling Pt: 83…84 °C
Melting Pt: 58…59 °C
Density: 1.021 g/cm³ (25 °C)
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00053515
CAS Number: 6263-65-6